Learn More
TWD Scientific LLC Large Polypropylene Weighing Funnels. Pack of 20
SDP
Catalog No. 501173373 Shop All TWD Scientific LLC Products
Encompass_Preferred
Disposable polypropylene weighing funnel, size large. Fits volumetric flask volumes (mL) of 500, 1000. Funnels have a maximum capacity of 20mL Solid/10mL Liquid. 20/Pack
This item is not returnable. View return policy
Catalog No. 50-117-3373 Supplier TWD Scientific LLC Supplier No. DPWFPP3LPK
Disposable polypropylene weighing funnel, size large. Fits volumetric flask volumes (mL) of 500, 1000. Funnels have a maximum capacity of 20mL Solid/10mL Liquid. 20/Pack