Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cayman Chemical L-DOPA | 59-92-7 | MFCD00002598 | 5 g
SDP

Supplier:  Cayman Chemical 132485

Encompass_Preferred

L-DOPA | Purity: 95% | Mol Wt: 197.2 | 59-92-7 | MFCD00002598 | 5 g

Catalog No. NC1822418


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.