Learn More
Cayman Chemical L-DOPA methyl ester (hydrochloride), Purity: ≥95%, CAS: 1421-65-4, 1 g
SDP
Catalog No. 502971690
Encompass_Preferred
A metabolic precursor of dopamine that crosses the blood brain barrier; used to treat Parkinson’s disease and stroke damage
Catalog No. 50-297-1690 Supplier Cayman Chemical Supplier No. 161491 G
A metabolic precursor of dopamine that crosses the blood brain barrier; used to treat Parkinson’s disease and stroke damage