Learn More
Medchemexpress LLC L-DOPA sodium 5mg
SDP
Catalog No. 5000669436
Encompass_Preferred
L-DOPA (Levodopa) sodium is an orally active metabolic precursor of neurotransmitters dopamine L-DOPA sodium can cross the blood-brain barrier and is converted into dopamine in the brain L-DOPA sodium has anti-allodynic effects and can be used for Parkinson s disease research[1][2][3]
Catalog No. 50-006-69436 Supplier Medchemexpress LLC Supplier No. 5MGHYN0304A
L-DOPA (Levodopa) sodium is an orally active metabolic precursor of neurotransmitters dopamine L-DOPA sodium can cross the blood-brain barrier and is converted into dopamine in the brain L-DOPA sodium has anti-allodynic effects and can be used for Parkinson s disease research[1][2][3]