Learn More
Medchemexpress LLC L-eflornithine monohydrochloride | 69955-42-6 | 98.0% | 218.63 | C6H13ClF2N2O2 | 10 MG
SDP
Catalog No. 5000297093
Encompass_Preferred
L-Eflornithine monohydrochloride is the L-enantiomer of eflornithine and an irreversible inhibitor of ornithine decarboxylase (ODC). It is provided as the monohydrochloride salt and is intended for laboratory research on polyamine metabolism, antiparasitic mechanisms, and related biochemical studies.
  • High chemical purity suitable for research (98.0%).
  • Monohydrochloride salt form for improved stability.
  • Irreversible ODC inhibitor used in polyamine metabolism studies.
  • Supplied in small quantities for assay development and pilot experiments.
  • Characterized by CAS 69955-42-6 and molecular weight 218.63.
Catalog No. 50-002-97093 Supplier Medchemexpress LLC Supplier No. HYB0744D10MG
L-Eflornithine monohydrochloride is the L-enantiomer of eflornithine and an irreversible inhibitor of ornithine decarboxylase (ODC). It is provided as the monohydrochloride salt and is intended for laboratory research on polyamine metabolism, antiparasitic mechanisms, and related biochemical studies.
  • High chemical purity suitable for research (98.0%).
  • Monohydrochloride salt form for improved stability.
  • Irreversible ODC inhibitor used in polyamine metabolism studies.
  • Supplied in small quantities for assay development and pilot experiments.
  • Characterized by CAS 69955-42-6 and molecular weight 218.63.