Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylpalladium chloride dimer, 98%

Catalog No. AC206830050
Change view
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC206830050 5 g Glass bottle
AC206835000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC206830050 Supplier Thermo Scientific Chemicals Supplier No. 206830050
Only null left
Add to Cart

CAS: 12012-95-2 | C6H10Cl2Pd2 | 365.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12012-95-2
Molecular Formula C6H10Cl2Pd2
Molecular Weight (g/mol) 365.89
MDL Number MFCD00044874
InChI Key PENAXHPKEVTBLF-UHFFFAOYSA-L
Synonym allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium
PubChem CID 61538
IUPAC Name chloropalladium(1+);prop-1-ene
SMILES Cl[Pd].Cl[Pd].c:c:c.c:c:c

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Green-Yellow to Yellow
Assay Percent Range 56.7% min. (Pd)
Packaging Glass bottle
Quantity 5 g
Fieser 02,109; 05,189; 07,5; 09,132
Solubility Information Solubility in water: decomposes
Formula Weight 365.84
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Allylpalladium chloride dimer, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 234-579-8
RTECSNumber RT3510000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.