Learn More
Cayman Chemical L-ErgothIonIn 10mg
SDP
Catalog No. 502973032
Encompass_Preferred
A naturally-occurring amino acid which is a stable antioxidant; scavenges free radicals and oxidants, controls NF-κB activation, and inhibits inflammatory gene expression; blocks oxidative DNA damage and cell death
Catalog No. 50-297-3032 Supplier Cayman Chemical Supplier No. 1490510 MG
A naturally-occurring amino acid which is a stable antioxidant; scavenges free radicals and oxidants, controls NF-κB activation, and inhibits inflammatory gene expression; blocks oxidative DNA damage and cell death