Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-erythro-2-pentulose | 2042-27-5 | >=96.0% | 25 MG
SDP

Catalog No. 5000231059
Encompass_Preferred

L-Ribulose solution is a 1 M aqueous solution of L-erythro-2-pentulose (CAS 2042-27-5) supplied as a small 25 mg quantity. It is used as a building block in rare sugar synthesis and L-nucleoside analogue preparation.

  • 1 M concentration in water.
  • Pack size provides approximately 166.5 μL at 1 M.
  • Purity >=96.0% by HPLC.
  • Molecular formula C5H10O5, molecular weight 150.13 g/mol.
  • Supplied with a certificate of analysis and product datasheet.
  • Useful precursor for synthesis of rare sugars and nucleoside analogues.

Catalog No. 50-002-31059 Supplier Medchemexpress LLC Supplier No. HY121305B25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Ribulose solution is a 1 M aqueous solution of L-erythro-2-pentulose (CAS 2042-27-5) supplied as a small 25 mg quantity. It is used as a building block in rare sugar synthesis and L-nucleoside analogue preparation.

  • 1 M concentration in water.
  • Pack size provides approximately 166.5 μL at 1 M.
  • Purity >=96.0% by HPLC.
  • Molecular formula C5H10O5, molecular weight 150.13 g/mol.
  • Supplied with a certificate of analysis and product datasheet.
  • Useful precursor for synthesis of rare sugars and nucleoside analogues.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.