Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-erythro-2-pentulose | 2042-27-5 | ≥96.0% | 5 MG
SDP

Catalog No. 5000232543
Encompass_Preferred

L-ribulose (L-erythro-2-pentulose) is the L-enantiomer of ribulose, used as a precursor for the synthesis of rare sugars and L-nucleoside analogues. Supplied as a small-mass reagent for laboratory research; not for human or clinical use.

  • L-enantiomer of ribulose for chemical synthesis
  • Molecular formula C5H10O5, molecular weight 150.13 g·mol⁻1
  • Purity ≥96.0% (certificate of analysis)
  • Available in small quantities suitable for synthetic and analytical work
  • Intended for research use only

Catalog No. 50-002-32543 Supplier Medchemexpress LLC Supplier No. HY121305A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-ribulose (L-erythro-2-pentulose) is the L-enantiomer of ribulose, used as a precursor for the synthesis of rare sugars and L-nucleoside analogues. Supplied as a small-mass reagent for laboratory research; not for human or clinical use.

  • L-enantiomer of ribulose for chemical synthesis
  • Molecular formula C5H10O5, molecular weight 150.13 g·mol⁻1
  • Purity ≥96.0% (certificate of analysis)
  • Available in small quantities suitable for synthetic and analytical work
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.