Learn More
Enzo Life Sciences L-erythro MAPP (25 mg). CAS: 143492-39-1
SDP
Catalog No. 502012134
Encompass_Preferred
Useful as a negative control for D-erythro-MAPP. It does not raise endogenous ceramide levels or inhibit the growth of HL-60 cells. Alternative name: (1S,2R-L-erythro-2-N-Myristoylamino)-1-phenyl-1-propanol. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2134 Supplier Enzo Life Sciences Supplier No. BMLSL2220025
Useful as a negative control for D-erythro-MAPP. It does not raise endogenous ceramide levels or inhibit the growth of HL-60 cells. Alternative name: (1S,2R-L-erythro-2-N-Myristoylamino)-1-phenyl-1-propanol. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml). Long Term Storage: -20°C.