Learn More
Cayman Chemical L-(+)-Erythrose, Purity: ≥95%, CAS: 533-49-3, 10 mg
SDP
Catalog No. 502985234
Encompass_Preferred
An aldotetrose carbohydrate that has been used in glycation studies and to characterize erythrose reductase activity
Catalog No. 50-298-5234 Supplier Cayman Chemical Supplier No. 2038110 MG
An aldotetrose carbohydrate that has been used in glycation studies and to characterize erythrose reductase activity