Learn More
Bioworld Lycopersicon esculentum (Tomato) Lectin (LEL/LEA) - Rhodamine (TRITC)
Catalog No. 50199142
Encompass
Lycopersicon esculentum (LEA) lectin purified from tomato fruit conjugated to Rhodamine (TRITC).Technical Specifications:Ligand Source:Lycopersicon esculentumMolecular Weight:71 kDaCarbohydrate Specificity:(GlcNAcβ4)n , (Galβ4GlcNAc)nInhibiting/Eluting Sugar:Chitin hydrolysateCarbohydrate for Elution:0.2MChitin hydrolysateBlood Group Specificity:N/AConjugate: RhodamineForm:LiquidIsoelectric (pI):>9Storage: -20°CApplication note: N/A
Catalog No. 50-199-142 Supplier Bioworld Supplier No. 217610312
Lycopersicon esculentum (LEA) lectin purified from tomato fruit conjugated to Rhodamine (TRITC).Technical Specifications:Ligand Source:Lycopersicon esculentumMolecular Weight:71 kDaCarbohydrate Specificity:(GlcNAcβ4)n , (Galβ4GlcNAc)nInhibiting/Eluting Sugar:Chitin hydrolysateCarbohydrate for Elution:0.2MChitin hydrolysateBlood Group Specificity:N/AConjugate: RhodamineForm:LiquidIsoelectric (pI):>9Storage: -20°CApplication note: N/A