Learn More
Bellco Glass, Inc. Large Flange Air Stone Sparger,15L
SDP
Supplier:  Bellco Glass, Inc. 196488156
Encompass_Preferred
Large flange air stone sparger for large flange vessels. Fits through stainless steel head plate fitting. These spargers fit through a 5/16" fitting. MADE IN THE USA. 316 l stainless steel Ra 32 finish or better Acrodisc filter assembly, 0.2 micron
Catalog No. 50-194-4886