Learn More
Bellco Glass, Inc. Large Flange Air Stone Sparger,8L
SDP
Catalog No. 501944882
Encompass_Preferred
Large flange air stone sparger for large flange vessels. Fits through stainless steel head plate fitting. These spargers fit through a 5/16" fitting. MADE IN THE USA. 316 l stainless steel Ra 32 finish or better Acrodisc filter assembly, 0.2 micron
Catalog No. 50-194-4882 Supplier Bellco Glass, Inc. Supplier No. 196488086
Large flange air stone sparger for large flange vessels. Fits through stainless steel head plate fitting. These spargers fit through a 5/16" fitting. MADE IN THE USA. 316 l stainless steel Ra 32 finish or better Acrodisc filter assembly, 0.2 micron