Learn More
Bellco Glass, Inc. L/F Drive Adapter, High Speed
SDP
Catalog No. 501946197
Encompass_Preferred
316 L Stainless Steel Large Flange Drive Adapter for use with High Speed Drive Adapter. For attachment of Overhead Drive Motors. Heavy duty cap assembly for large flange vessels Adapt overhead drives to teflon or sst folding impeller assembly 316 sst design with polyester stainless steel bearings. MADE IN THE USA
Catalog No. 50-194-6197 Supplier Bellco Glass, Inc. Supplier No. 776472000
316 L Stainless Steel Large Flange Drive Adapter for use with High Speed Drive Adapter. For attachment of Overhead Drive Motors. Heavy duty cap assembly for large flange vessels Adapt overhead drives to teflon or sst folding impeller assembly 316 sst design with polyester stainless steel bearings. MADE IN THE USA