Learn More
Bellco Glass, Inc. Large Flange Flat Bottom Omni Vessel,36L
SDP
Catalog No. 501944888
Encompass_Preferred
Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp.Can be used with series 500 controllers or with CellTrol II (no speed) and OHD or high speed motor system. Large flange omni vessels for use with stainless steel headplate, 5l thru 36l. MADE IN THE USA. Heavy walled construction for cell culture and fermentation applicationsHeavy duty o-ring flange creates positive sealFor use with stainless steel headplate, o-ring and locking clamp36L flat bottom omni vesselOD = 410mm , H = 515mmUse with 200mm large flange headplate
Catalog No. 50-194-4888 Supplier Bellco Glass, Inc. Supplier No. 196488360
Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp.Can be used with series 500 controllers or with CellTrol II (no speed) and OHD or high speed motor system. Large flange omni vessels for use with stainless steel headplate, 5l thru 36l. MADE IN THE USA. Heavy walled construction for cell culture and fermentation applicationsHeavy duty o-ring flange creates positive sealFor use with stainless steel headplate, o-ring and locking clamp36L flat bottom omni vesselOD = 410mm , H = 515mmUse with 200mm large flange headplate