Learn More
Bellco Glass, Inc. Large Flange Omni-Paddle,8L
SDP
Catalog No. 501944881
Encompass_Preferred
Blades and impeller assemblies are use for cell culture. Reduces shear in delicate cell culture. Manufactured from 316l stainless steel with electropolish finish. Ra 32 finish or better inside and out. MADE IN THE USA. Blade dimension: 5 3/4 x 2". Shaft oal: 12 1/2".
Catalog No. 50-194-4881 Supplier Bellco Glass, Inc. Supplier No. 196488082
Blades and impeller assemblies are use for cell culture. Reduces shear in delicate cell culture. Manufactured from 316l stainless steel with electropolish finish. Ra 32 finish or better inside and out. MADE IN THE USA. Blade dimension: 5 3/4 x 2". Shaft oal: 12 1/2".