Learn More
Bellco Glass, Inc. Large Flange Round Bottom Omni Vessel,15L
SDP
Catalog No. 501944895
Encompass_Preferred
15L round bottom omni vessel 292 x 500mm Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Use with series 500 controllers or with CellTrol (no speed) and OHD or high speed motor system. Use with 200mm large flange headplate. MADE IN THE USA.
Catalog No. 50-194-4895 Supplier Bellco Glass, Inc. Supplier No. 196488415
15L round bottom omni vessel 292 x 500mm Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Use with series 500 controllers or with CellTrol (no speed) and OHD or high speed motor system. Use with 200mm large flange headplate. MADE IN THE USA.