Learn More
Bellco Glass, Inc. Large Flange Round Bottom Omni Vessel,36L
SDP
Catalog No. 501944896
Encompass_Preferred
36L round bottom omni vessel 410 x 550mm. Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Use with series 500 controllers or CellTrol (no speed) and OHD or high speed motor system. Takes 200mm large flange headplate. MADE IN THE USA.
Catalog No. 50-194-4896 Supplier Bellco Glass, Inc. Supplier No. 196488436
36L round bottom omni vessel 410 x 550mm. Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Use with series 500 controllers or CellTrol (no speed) and OHD or high speed motor system. Takes 200mm large flange headplate. MADE IN THE USA.