Learn More
Bellco Glass, Inc. Large Flange Round Bottom Omni Vessel,8L
SDP
Catalog No. 501944893
Encompass_Preferred
8L omni vessel 238 x 445mm, round bottom. Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Use with 200mm large flange headplate. Use with series 500 controllers or with CellTrol (no speed) and OHD or high speed motor system. MADE IN THE USA.
Catalog No. 50-194-4893 Supplier Bellco Glass, Inc. Supplier No. 196488408
8L omni vessel 238 x 445mm, round bottom. Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Use with 200mm large flange headplate. Use with series 500 controllers or with CellTrol (no speed) and OHD or high speed motor system. MADE IN THE USA.