Learn More
Bellco Glass, Inc. Large Flange Round Bottom Vessel,3L
SDP
Catalog No. 501944891
Encompass_Preferred
3L bioreactor vessel 145 x 250mm, round bottom. Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Use with CellTrol II only single or dual system and speed control modules. MADE IN THE USA.
Catalog No. 50-194-4891 Supplier Bellco Glass, Inc. Supplier No. 196488403
3L bioreactor vessel 145 x 250mm, round bottom. Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Use with CellTrol II only single or dual system and speed control modules. MADE IN THE USA.