Learn More
Bellco Glass, Inc. Large Flange Round Bottom Vessel,7L
SDP
Catalog No. 501944892
Encompass_Preferred
7L bioreactor vessel 200 x 390mm, round bottom. Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Use with Cell Trol II only and speed control module. MADE IN THE USA.
Catalog No. 50-194-4892 Supplier Bellco Glass, Inc. Supplier No. 196488407
7L bioreactor vessel 200 x 390mm, round bottom. Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. Use with Cell Trol II only and speed control module. MADE IN THE USA.