Learn More
Bellco Glass, Inc. Large Flange Stainless Steel Clamp, 8 OD "
SDP
Catalog No. 501944904
Encompass_Preferred
200 mm SST large flange headplate clamp fits headplates, 5 liter thru 36 liter vessels. MADE IN THE USA. Corrosion resistant 316l stainless steel Includes red silicone o-ring and 200mm t-bolt clamp
Catalog No. 50-194-4904 Supplier Bellco Glass, Inc. Supplier No. 196488700
200 mm SST large flange headplate clamp fits headplates, 5 liter thru 36 liter vessels. MADE IN THE USA. Corrosion resistant 316l stainless steel Includes red silicone o-ring and 200mm t-bolt clamp