Learn More
Bellco Glass, Inc. Large Flange Z/Z Paddle,15L
SDP
Catalog No. 501944887
Encompass_Preferred
Manufactured from 316l stainless steel with electropolish finish. Ra 32 finish or better inside and out. Ideal use for cell culture and reduces shear in delicate cell culture. MADE IN THE USA. Blade dimension: 5 3/4 x 2 1/2" Shaft oal: 14 1/4" Constructed of 316L ELECTROPOLISHED STAINLESS STEEL MARINE IMPELLER INCLUDES 1/2" SHAFT AND IMPELLER BLADE OMNI AND Z-Z PADDLES ARE ONE PIECE WELDED, 1/4" SHAFT
Catalog No. 50-194-4887 Supplier Bellco Glass, Inc. Supplier No. 196488158
Manufactured from 316l stainless steel with electropolish finish. Ra 32 finish or better inside and out. Ideal use for cell culture and reduces shear in delicate cell culture. MADE IN THE USA. Blade dimension: 5 3/4 x 2 1/2" Shaft oal: 14 1/4" Constructed of 316L ELECTROPOLISHED STAINLESS STEEL MARINE IMPELLER INCLUDES 1/2" SHAFT AND IMPELLER BLADE OMNI AND Z-Z PADDLES ARE ONE PIECE WELDED, 1/4" SHAFT