Learn More
Bellco Glass, Inc. Large Flange Z/Z Paddle, 36L
SDP
Catalog No. 501944890
Encompass_Preferred
Manufactured from 316l stainless steel with electropolish finish. Ra 32 finish or better inside and out. Ideal use for cell culture and reduces shear in delicate cell culture. MADE IN THE USA. Blade dimension: 5 3/4 x 2 1/2" Shaft oal: 16 3/4"
Catalog No. 50-194-4890 Supplier Bellco Glass, Inc. Supplier No. 196488368
Manufactured from 316l stainless steel with electropolish finish. Ra 32 finish or better inside and out. Ideal use for cell culture and reduces shear in delicate cell culture. MADE IN THE USA. Blade dimension: 5 3/4 x 2 1/2" Shaft oal: 16 3/4"