Learn More
Bellco Glass, Inc. Large Flange Z/Z Paddle,5L
SDP
Catalog No. 501944879
Encompass_Preferred
Blades and impeller assemblies are use for cell culture. Reduces shear in delicate cell culture. Manufactured from 316l stainless steel with electropolish finish. Ra 32 finish or better inside and out. MADE IN THE USA. Blade dimension: 5 3/4 x 2 1/2". Shaft oal: 6 1/2". MARINE IMPELLER INCLUDES 1/2" SHAFT AND IMPELLER BLADE OMNI AND Z-Z PADDLES ARE ONE PIECE WELDED, 1/4" SHAFT
Catalog No. 50-194-4879 Supplier Bellco Glass, Inc. Supplier No. 196488058
Blades and impeller assemblies are use for cell culture. Reduces shear in delicate cell culture. Manufactured from 316l stainless steel with electropolish finish. Ra 32 finish or better inside and out. MADE IN THE USA. Blade dimension: 5 3/4 x 2 1/2". Shaft oal: 6 1/2". MARINE IMPELLER INCLUDES 1/2" SHAFT AND IMPELLER BLADE OMNI AND Z-Z PADDLES ARE ONE PIECE WELDED, 1/4" SHAFT