Learn More
Medchemexpress LLC L-(+)-Fructose | 7776-48-9 | 100 MG

Supplier: Medchemexpress LLC HYN7092A100MG
L-(+)-Fructose is the L-isomer of D-Fructose. It is involved in glycolysis and gluconeogenesis, and can be used to study photosynthesis and carbohydrate storage in plant biology. It can also be used to investigate the activity of fructokinase and other fructose-metabolizing enzymes.
- Involved in glycolysis and gluconeogenesis
- Can be used to study photosynthesis and carbohydrate storage in plant biology
- Can be used to investigate the activity of fructokinase and other fructose-metabolizing enzymes
- Molecular weight: 180.16
- Formula: C6H12O6
- Appearance: White to off-white solid
- Structure classified as saccharides
- Initial source: Plants Elaeagnaceae Elaeagnus angustifolia L.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.