Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-(+)-Fructose | 7776-48-9 | 25 MG
SDP

Catalog No. 5000421611
Encompass_Preferred

L-(+)-Fructose is the L-isomer of D-Fructose. It is involved in glycolysis and gluconeogenesis, and can be utilized to study photosynthesis and carbohydrate storage in plant biology. It can also be used to investigate the activity of fructokinase and other fructose-metabolizing enzymes.

  • Purity of 95.0%
  • Molecular weight of 180.16
  • Chemical formula C6H12O6
  • Appears as a solid
  • Color white to off-white
  • Classified as a saccharide
  • Derived from plants Elaeagnaceae Elaeagnus angustifolia L.
  • Soluble in water at 116.67 mg/mL

Catalog No. 50-004-21611 Supplier Medchemexpress LLC Supplier No. 25MGHYN7092A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-(+)-Fructose is the L-isomer of D-Fructose. It is involved in glycolysis and gluconeogenesis, and can be utilized to study photosynthesis and carbohydrate storage in plant biology. It can also be used to investigate the activity of fructokinase and other fructose-metabolizing enzymes.

  • Purity of 95.0%
  • Molecular weight of 180.16
  • Chemical formula C6H12O6
  • Appears as a solid
  • Color white to off-white
  • Classified as a saccharide
  • Derived from plants Elaeagnaceae Elaeagnus angustifolia L.
  • Soluble in water at 116.67 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.