Learn More
Medchemexpress LLC L-(+)-Fructose | 7776-48-9 | 50 MG
SDP
Catalog No. 5000419676
Encompass_Preferred
L-(+)-Fructose is the L-isomer of D-Fructose. It is involved in glycolysis and gluconeogenesis. This product can be utilized for studying photosynthesis and carbohydrate storage in plant biology. Additionally, it can be used to investigate the activity of fructokinase and other fructose-metabolizing enzymes.
  • Involved in glycolysis and gluconeogenesis.
  • Utilized for studying photosynthesis and carbohydrate storage in plant biology.
  • Used to investigate the activity of fructokinase and other fructose-metabolizing enzymes.
Catalog No. 50-004-19676 Supplier Medchemexpress LLC Supplier No. HYN7092A50MG
L-(+)-Fructose is the L-isomer of D-Fructose. It is involved in glycolysis and gluconeogenesis. This product can be utilized for studying photosynthesis and carbohydrate storage in plant biology. Additionally, it can be used to investigate the activity of fructokinase and other fructose-metabolizing enzymes.
  • Involved in glycolysis and gluconeogenesis.
  • Utilized for studying photosynthesis and carbohydrate storage in plant biology.
  • Used to investigate the activity of fructokinase and other fructose-metabolizing enzymes.