Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-(+)-Fructose, 95% | 7776-48-9 | 10 MG
SDP

Catalog No. 5000418045
Encompass_Preferred

L-(+)-Fructose, 95% is the L-isomer of D-Fructose. It is involved in glycolysis and gluconeogenesis, and can be used to study photosynthesis and carbohydrate storage in plant biology. This chemical can also be used to investigate the activity of fructokinase and other fructose-metabolizing enzymes. It is for research use only.

  • L-isomer of D-Fructose
  • Involved in glycolysis and gluconeogenesis
  • Can be used to study photosynthesis and carbohydrate storage in plant biology
  • Can be used to investigate the activity of fructokinase and other fructose-metabolizing enzymes
  • Molecular weight: 180.16
  • Formula: C6H12O6
  • Appearance: White to off-white solid
  • Initial source: Plants Elaeagnaceae Elaeagnus angustifolia L.

Catalog No. 50-004-18045 Supplier Medchemexpress LLC Supplier No. HYN7092A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-(+)-Fructose, 95% is the L-isomer of D-Fructose. It is involved in glycolysis and gluconeogenesis, and can be used to study photosynthesis and carbohydrate storage in plant biology. This chemical can also be used to investigate the activity of fructokinase and other fructose-metabolizing enzymes. It is for research use only.

  • L-isomer of D-Fructose
  • Involved in glycolysis and gluconeogenesis
  • Can be used to study photosynthesis and carbohydrate storage in plant biology
  • Can be used to investigate the activity of fructokinase and other fructose-metabolizing enzymes
  • Molecular weight: 180.16
  • Formula: C6H12O6
  • Appearance: White to off-white solid
  • Initial source: Plants Elaeagnaceae Elaeagnus angustifolia L.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.