Learn More
Cayman Chemical L-Fucose 10g
SDP
Catalog No. 502971379
Encompass_Preferred
A deoxyhexose monosaccharide found on N- and O-linked glycans and glycolipids that exists either as a terminal modification of glycan structures or serves as a point of attachment for adding other sugars; plays a role in A and B blood group antigen substructure determination, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions
Catalog No. 50-297-1379 Supplier Cayman Chemical Supplier No. 1647910 G
Add to Cart
Add to Cart
A deoxyhexose monosaccharide found on N- and O-linked glycans and glycolipids that exists either as a terminal modification of glycan structures or serves as a point of attachment for adding other sugars; plays a role in A and B blood group antigen substructure determination, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions