Learn More
Cederlane L-Fucose Assay Kit
SDP
Catalog No. NC1817542
Encompass
The L-Fucose test kit is a simple, rapid and reliable method, for the measurement and analysis of L-Fucose in plant extracts, biological samples and other materials. This kit can be used in the measurement of α-fucosidases that do not act on chromogenic substrates.
Catalog No. NC1817542 Supplier Cederlane Supplier No. 700004288
The L-Fucose test kit is a simple, rapid and reliable method, for the measurement and analysis of L-Fucose in plant extracts, biological samples and other materials. This kit can be used in the measurement of α-fucosidases that do not act on chromogenic substrates.