Learn More
TWD Scientific LLC Large Glass Weighing Funnel. Pack of 6
SDP
Catalog No. 501173391 Shop All TWD Scientific LLC Products
Encompass_Preferred
Disposable borosilicate glass weighing funnel, size large. Fits volumetric flask volumes (mL) of 500, 1000. Funnels have a maximum capacity of 20mL Solid/10mL Liquid. 6/Pack
Catalog No. 50-117-3391 Supplier TWD Scientific LLC Supplier No. GWFBS3LPK
Disposable borosilicate glass weighing funnel, size large. Fits volumetric flask volumes (mL) of 500, 1000. Funnels have a maximum capacity of 20mL Solid/10mL Liquid. 6/Pack