Learn More
Medchemexpress LLC L-Glucose-13C | 478519-02-7 | 98.0% | 181.15 | 100 MG
SDP
Catalog No. 5000451554
Encompass_Preferred
L-Glucose-13C is the 13C labeled form of L-Glucose. L-Glucose (L-(-)-Glucose) is an enantiomer of D-glucose and has been shown to promote food intake. It is intended for research use only.
  • Labeled with 13C isotope
  • Enantiomer of D-glucose
  • Can promote food intake
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-51554 Supplier Medchemexpress LLC Supplier No. HYW010042S100MG
L-Glucose-13C is the 13C labeled form of L-Glucose. L-Glucose (L-(-)-Glucose) is an enantiomer of D-glucose and has been shown to promote food intake. It is intended for research use only.
  • Labeled with 13C isotope
  • Enantiomer of D-glucose
  • Can promote food intake
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS