Learn More
Medchemexpress LLC L-Glucose-13C | 478519-02-7 | 98.0% | 181.15 | 25 MG
SDP
Catalog No. 5000453244
Encompass_Preferred
L-Glucose-13C is the 13C labeled L-Glucose, an enantiomer of D-glucose. It can promote food intake and is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-53244 Supplier Medchemexpress LLC Supplier No. 25MGHYW010042S
L-Glucose-13C is the 13C labeled L-Glucose, an enantiomer of D-glucose. It can promote food intake and is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.