Learn More
Medchemexpress LLC L-Glucose-13C | 478519-02-7 | 98.0% | 181.15 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW010042S
Encompass_Preferred
L-Glucose-13C is the 13C labeled L-Glucose. L-Glucose (L-(-)-Glucose) is an enantiomer of D-glucose. It can promote food intake.
  • 13C labeled L-Glucose
  • Enantiomer of D-glucose
  • Can promote food intake
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-66349