Learn More
Medchemexpress LLC L-Glucose-13C | 478519-02-7 | 98.0% | 181.15 | 50 MG
SDP
Catalog No. 5000453536
Encompass_Preferred
L-Glucose-13C is the 13C labeled L-Glucose, an enantiomer of D-glucose. It is for research use only and can promote food intake. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-53536 Supplier Medchemexpress LLC Supplier No. HYW010042S50MG
L-Glucose-13C is the 13C labeled L-Glucose, an enantiomer of D-glucose. It is for research use only and can promote food intake. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS