Learn More
Medchemexpress LLC L-Glucose-13C6 | 921-60-8 | 99.4% | 186.11 | 1 MG
SDP
Catalog No. 5000366427
Encompass_Preferred
L-Glucose-13C6 is C13-labeled L-Glucose, an enantiomer of D-glucose that enhances food intake. It is largely used as a tracer for quantitation during the drug development process due to the incorporation of stable heavy isotopes. It can also serve as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-66427 Supplier Medchemexpress LLC Supplier No. HYW010042S31MG
L-Glucose-13C6 is C13-labeled L-Glucose, an enantiomer of D-glucose that enhances food intake. It is largely used as a tracer for quantitation during the drug development process due to the incorporation of stable heavy isotopes. It can also serve as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS