Learn More
Cayman Chemical L-(−)-Glucose, Purity: ≥95%, CAS: 921-60-8, 1 g
SDP
Catalog No. 502981284
Encompass_Preferred
An enantiomer D-glucose that is not bioavailable to cells as an energy source because it cannot be phosphorylated by hexokinase
Catalog No. 50-298-1284 Supplier Cayman Chemical Supplier No. 208291 G
An enantiomer D-glucose that is not bioavailable to cells as an energy source because it cannot be phosphorylated by hexokinase