Learn More
Cayman Chemical L-(−)-Glucose, Purity: ≥95%, CAS: 921-60-8, 250 mg
SDP
Catalog No. 502981330
Encompass_Preferred
An enantiomer D-glucose that is not bioavailable to cells as an energy source because it cannot be phosphorylated by hexokinase
Catalog No. 50-298-1330 Supplier Cayman Chemical Supplier No. 20829250 MG
An enantiomer D-glucose that is not bioavailable to cells as an energy source because it cannot be phosphorylated by hexokinase