Learn More
Medchemexpress LLC L-Glucose | 921-60-8 | 99.89% | 180.16 | 25 G
SDP
Catalog No. 5000452406
Encompass_Preferred
L-Glucose is a stereoisomer of D-Glucose that does not readily enter the brain. It can promote food intake and is used in research to enhance memory in mice. When combined with a fluorescence detector, L-Glucose can produce a fluorescent probe for visualizing and characterizing cancer cells. This product is for research use only and not sold to patients.
  • Promotes food intake
  • Used to enhance memory in mice
  • Can be combined with a fluorescence detector to create a fluorescent probe for visualizing and characterizing cancer cells
Catalog No. 50-004-52406 Supplier Medchemexpress LLC Supplier No. 25GHYW010042
L-Glucose is a stereoisomer of D-Glucose that does not readily enter the brain. It can promote food intake and is used in research to enhance memory in mice. When combined with a fluorescence detector, L-Glucose can produce a fluorescent probe for visualizing and characterizing cancer cells. This product is for research use only and not sold to patients.
  • Promotes food intake
  • Used to enhance memory in mice
  • Can be combined with a fluorescence detector to create a fluorescent probe for visualizing and characterizing cancer cells