Learn More
Medchemexpress LLC L-(-)-Glucose | 921-60-8 | 99.89% | 180.16 | 500 MG
SDP
Catalog No. 5000453637
Encompass_Preferred
L-Glucose is a stereoisomer of D-Glucose that does not readily enter the brain. It can promote food intake and can be combined with a fluorescence detector to produce a fluorescent probe for visualizing and characterizing cancer cells. L-Glucose can also be used in research to enhance memory in mice.
  • Does not readily enter the brain
  • Can promote food intake
  • Can be combined with a fluorescence detector to produce a fluorescent probe for visualizing and characterizing cancer cells
  • Can be used in research to enhance memory in mice
Catalog No. 50-004-53637 Supplier Medchemexpress LLC Supplier No. HYW010042500MG
L-Glucose is a stereoisomer of D-Glucose that does not readily enter the brain. It can promote food intake and can be combined with a fluorescence detector to produce a fluorescent probe for visualizing and characterizing cancer cells. L-Glucose can also be used in research to enhance memory in mice.
  • Does not readily enter the brain
  • Can promote food intake
  • Can be combined with a fluorescence detector to produce a fluorescent probe for visualizing and characterizing cancer cells
  • Can be used in research to enhance memory in mice