Learn More
Medchemexpress LLC L-Glucose | 921-60-8 | 99.9% | 180.16 | 1 G
SDP
Catalog No. 5000453535
Encompass_Preferred
L-Glucose is a stereoisomer of D-Glucose that does not readily enter the brain. It can promote food intake and can be combined with a fluorescence detector to produce a fluorescent probe for visualizing and characterizing cancer cells. L-Glucose also can be used in research to enhance memory in mice.
  • Does not readily enter the brain.
  • Can promote food intake.
  • Can be combined with a fluorescence detector to produce a fluorescent probe for visualizing and characterizing cancer cells.
  • Can be used to enhance memory in mice.
Catalog No. 50-004-53535 Supplier Medchemexpress LLC Supplier No. HYW0100421G
L-Glucose is a stereoisomer of D-Glucose that does not readily enter the brain. It can promote food intake and can be combined with a fluorescence detector to produce a fluorescent probe for visualizing and characterizing cancer cells. L-Glucose also can be used in research to enhance memory in mice.
  • Does not readily enter the brain.
  • Can promote food intake.
  • Can be combined with a fluorescence detector to produce a fluorescent probe for visualizing and characterizing cancer cells.
  • Can be used to enhance memory in mice.