Learn More
Medchemexpress LLC L-Glucose | 921-60-8 | 99.9% | 180.16 | 1 ML
SDP
Catalog No. 5000453941
Encompass_Preferred
L-Glucose (L-(-)-Glucose) is a stereoisomer of D-Glucose that does not readily enter the brain. It can promote food intake and, when combined with a fluorescence detector, can produce a fluorescent probe for visualizing and characterizing cancer cells. It can also be used in research to enhance memory in mice.
  • Stereoisomer of D-glucose.
  • Does not readily enter the brain.
  • Can promote food intake.
  • Can be combined with a fluorescence detector to produce a fluorescent probe for visualizing and characterizing cancer cells.
  • Can enhance memory in mice.
  • Appearance: Solid.
  • Color: White to off-white.
Catalog No. 50-004-53941 Supplier Medchemexpress LLC Supplier No. HYW01004210MM/1ML
L-Glucose (L-(-)-Glucose) is a stereoisomer of D-Glucose that does not readily enter the brain. It can promote food intake and, when combined with a fluorescence detector, can produce a fluorescent probe for visualizing and characterizing cancer cells. It can also be used in research to enhance memory in mice.
  • Stereoisomer of D-glucose.
  • Does not readily enter the brain.
  • Can promote food intake.
  • Can be combined with a fluorescence detector to produce a fluorescent probe for visualizing and characterizing cancer cells.
  • Can enhance memory in mice.
  • Appearance: Solid.
  • Color: White to off-white.