Learn More
Medchemexpress LLC L-Glucose | 921-60-8 | 99.9% | 180.16 | 5 G
SDP
Catalog No. 5000452501
Encompass_Preferred
L-Glucose (L-(-)-Glucose) is a stereoisomer of D-Glucose that does not readily enter the brain. It can promote food intake and may also be used in research to enhance memory in mice.
  • Can be combined with a fluorescence detector to create a fluorescent probe for visualizing and characterizing cancer cells.
  • Enhances place learning and decreases escape latencies in mice.
Catalog No. 50-004-52501 Supplier Medchemexpress LLC Supplier No. HYW0100425G
L-Glucose (L-(-)-Glucose) is a stereoisomer of D-Glucose that does not readily enter the brain. It can promote food intake and may also be used in research to enhance memory in mice.
  • Can be combined with a fluorescence detector to create a fluorescent probe for visualizing and characterizing cancer cells.
  • Enhances place learning and decreases escape latencies in mice.