Learn More
American Radiolabeled Chemicals Inc L-GLUT 5-14C 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 356250UC
Encompass_Preferred
Glutamine, L-5-14C 50uCi in sterile water, 0.1 mCi/ml, 50-60 mCi/mmol, M.W. 146.2, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-946
Add to Cart
Request a Quote