Learn More
Cosmo Bio Usa Inc L GLUTAMATE OXIDASE 25 RECOMB
SDP
Catalog No. NC0302998
Encompass
Recombinant L-Glutamate Oxidase (EC 1.4.3.11) produced in E. Coli. Lyophilized. 25 units/vial. Does not react with glutamine, aspartate, D-glutamate or 16 other L-amino acids.
Catalog No. NC0302998 Supplier Cosmo Bio Usa Inc Supplier No. YMS80049
Recombinant L-Glutamate Oxidase (EC 1.4.3.11) produced in E. Coli. Lyophilized. 25 units/vial. Does not react with glutamine, aspartate, D-glutamate or 16 other L-amino acids.