Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Glutamate Oxidase | CAS 9068-41-1 | 5 units
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G19245UN
Encompass
L-glutamate oxidase | Purity: N/A | Mol Wt: 250 kDa | CAS No: 9068-41-1 | EC No: 1.4.3.11 | 5 units
Catalog No. NC2603465