Learn More
Medchemexpress LLC L-glutamic-2,4,4-d3 acid | 203805-84-9 | 98.0% | 150.15 g/mol | C5H6D3NO4 | 5 MG
SDP
Catalog No. 5000222291
Encompass_Preferred
L-Glutamic-2,4,4-d3 acid is a deuterium-labeled analogue of L-glutamic acid provided as a research-grade stable isotope standard. It is used in tracer studies, quantitative mass spectrometry, and analytical method development where isotopic enrichment is required. The product is supplied as a high-purity solid with batch COA and SDS available for verification.
  • Deuterium-labeled analogue for isotopic tracing.
  • Suitable for quantitative mass spectrometry and method development.
  • High purity (98.0%) appropriate for analytical applications.
  • Supplied as a solid in small research quantities for convenient handling.
  • Batch COA and SDS available for quality and safety verification.
Catalog No. 50-002-22291 Supplier Medchemexpress LLC Supplier No. HY14608S85MG
L-Glutamic-2,4,4-d3 acid is a deuterium-labeled analogue of L-glutamic acid provided as a research-grade stable isotope standard. It is used in tracer studies, quantitative mass spectrometry, and analytical method development where isotopic enrichment is required. The product is supplied as a high-purity solid with batch COA and SDS available for verification.
  • Deuterium-labeled analogue for isotopic tracing.
  • Suitable for quantitative mass spectrometry and method development.
  • High purity (98.0%) appropriate for analytical applications.
  • Supplied as a solid in small research quantities for convenient handling.
  • Batch COA and SDS available for quality and safety verification.