Learn More
Medchemexpress LLC L-glutamic-2,4,4-d3 acid | 203805-84-9 | 98.0% | C5H6D3NO4 | 10MG
SDP
Catalog No. 5000258392
Encompass_Preferred
L-Glutamic acid-d3 is a deuterium-labeled form of L-glutamic acid used as a stable-isotope reference for analytical and biochemical workflows. It is provided as a high-purity small-quantity powder suitable for use as an internal standard in mass spectrometry, NMR spectroscopy, and metabolic tracing. The material has molecular formula C5H6D3NO4 and is supplied with quality documentation; recommended storage is -20°C under nitrogen, and solutions should be kept at -80°C for long-term stability.
  • Deuterium-labeled analogue for isotope tracing and internal standard applications.
  • High purity (98.0%) appropriate for quantitative analysis.
  • Compatible with mass spectrometry and NMR workflows.
  • Stable when stored at -20°C under inert atmosphere; in solution store at -80°C for extended shelf life.
  • Available in small milligram pack sizes for analytical use.
Catalog No. 50-002-58392 Supplier Medchemexpress LLC Supplier No. HY14608S810MG
L-Glutamic acid-d3 is a deuterium-labeled form of L-glutamic acid used as a stable-isotope reference for analytical and biochemical workflows. It is provided as a high-purity small-quantity powder suitable for use as an internal standard in mass spectrometry, NMR spectroscopy, and metabolic tracing. The material has molecular formula C5H6D3NO4 and is supplied with quality documentation; recommended storage is -20°C under nitrogen, and solutions should be kept at -80°C for long-term stability.
  • Deuterium-labeled analogue for isotope tracing and internal standard applications.
  • High purity (98.0%) appropriate for quantitative analysis.
  • Compatible with mass spectrometry and NMR workflows.
  • Stable when stored at -20°C under inert atmosphere; in solution store at -80°C for extended shelf life.
  • Available in small milligram pack sizes for analytical use.